Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNKS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025235.3 | NP_079511.1 | 16, 17 | 2350 | ||
XM_005270185.4 | XP_005270242.2 | 16, 17 | 2390 | |||
XM_011540213.1 | XP_011538515.1 | 16, 17 | 2381 | |||
XM_011540215.2 | XP_011538517.1 | 8, 9 | 2575 | |||
XM_017016696.1 | XP_016872185.1 | 16, 17 | 2327 | |||
XM_017016697.1 | XP_016872186.1 | 16, 17 | 2343 | |||
XM_017016698.1 | XP_016872187.1 | 16, 17 | 2054 | |||
XM_017016699.1 | XP_016872188.1 | 16, 17 | 2334 | |||
XM_017016700.1 | XP_016872189.1 | 16, 17 | 1940 | |||
GenBank mRNA | AF264912.1 | - | 16, 17 | 2053 | ||
AF305081.1 | - | 16, 17 | 2352 | |||
AF309033.1 | - | 16, 17 | 2304 | |||
AF329696.1 | - | 16, 17 | 2192 | |||
AF342982.1 | - | 16, 17 | 2053 | |||
AF438201.1 | - | 16, 17 | 2432 | |||
AK314612.1 | - | 16, 17 | 2318 | |||
AL833676.1 | - | 2, 3 | 388 |