Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025245.2 | NP_079521.1 | 3 | 699 | ||
XM_006722911.3 | XP_006722974.1 | 2 | 280 | |||
XM_011528320.2 | XP_011526622.1 | 4 | 745 | |||
XM_011528322.2 | XP_011526624.1 | 2 | 334 | |||
XM_017027326.1 | XP_016882815.1 | 4 | 380 | |||
XM_017027327.1 | XP_016882816.1 | 4 | 361 | |||
XM_017027328.1 | XP_016882817.1 | 4 | 745 | |||
GenBank mRNA | AJ300182.1 | - | 2 | 280 | ||
AK097427.1 | - | 3 | 436 | |||
BC141859.1 | - | 3 | 699 | 8860384 |