Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030636.2 | NP_085139.2 | 8 | 2188 | ||
GenBank mRNA | AB051493.1 | - | 8 | 1929 | ||
AK027386.1 | - | 8 | 2160 | |||
BC065518.1 | - | 9 | 2088 | 5755727 |