Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306199.1 | NP_001293128.1 | 4 | 1018 | ||
NM_030648.3 | NP_085151.1 | 4 | 1018 | |||
XM_017008661.1 | XP_016864150.1 | 2 | 111 | |||
GenBank mRNA | AB051504.1 | - | 4 | 653 | ||
AF448510.1 | - | 4 | 653 | |||
AF462150.1 | - | 4 | 386 | |||
AK310856.1 | - | 4 | 487 | |||
BC121055.1 | - | 4 | 562 | 40118363 | ||
BC121056.1 | - | 4 | 562 | 40118365 |