Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPRSS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288749.1 | NP_001275678.1 | 5 | 536 | ||
NM_001288750.1 | NP_001275679.1 | 5 | 536 | |||
NM_001288751.1 | NP_001275680.1 | 6 | 743 | |||
NM_001288752.1 | NP_001275681.1 | 6 | 639 | |||
NM_030770.3 | NP_110397.2 | 6 | 639 | |||
NR_110046.1 | 4 | 437 | ||||
NR_110047.1 | 4 | 437 | ||||
XM_017018366.1 | XP_016873855.1 | 4 | 3875 | |||
XM_017018367.1 | XP_016873856.1 | 6 | 4067 | |||
GenBank mRNA | AB028140.1 | - | 6 | 645 | ||
AK294313.1 | - | 5 | 471 | |||
AK295366.1 | - | 6 | 568 | |||
AL538140.3 | - | 6 | 580 | |||
BC121802.1 | - | 5 | 509 | 40085103 | ||
BC121803.1 | - | 6 | 716 | 40085104 |