Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030780.4 | NP_110407.2 | 2 | 571 | ||
NR_102337.1 | 2 | 571 | ||||
NR_102338.1 | 3 | 680 | ||||
XM_017013877.1 | XP_016869366.1 | 3 | 560 | |||
GenBank mRNA | AF283645.1 | - | 2 | 301 | ||
AK027531.1 | - | 2 | 340 | |||
AK027787.1 | - | 2 | 305 | |||
AK125492.1 | - | 3 | 289 | |||
AK300748.1 | - | 2 | 305 | |||
AL046268.1 | - | 2 | 141 | |||
BC021893.1 | - | 2 | 279 | 4524907 | ||
BP274577.1 | - | 2 | 476 | |||
DC331449.1 | - | 2 | 69 |