Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030780.4 | NP_110407.2 | 5 | 997 | ||
NR_102337.1 | 4 | 911 | ||||
NR_102338.1 | 6 | 1106 | ||||
XM_017013877.1 | XP_016869366.1 | 5 | 900 | |||
GenBank mRNA | AF283645.1 | - | 5 | 727 | ||
AK027531.1 | - | 5 | 766 | |||
AK027787.1 | - | 5 | 731 | |||
AK125492.1 | - | 6 | 715 | |||
AK300748.1 | - | 4 | 645 | |||
BC021893.1 | - | 5 | 705 | 4524907 | ||
DC331449.1 | - | 4 | 409 |