Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFHR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030787.3 | NP_110414.1 | 4 | 708 | ||
XM_011510020.2 | XP_011508322.1 | 4 | 615 | |||
GenBank mRNA | AF295327.1 | - | 4 | 673 | ||
AK314026.1 | - | 4 | 709 | |||
BC026282.1 | - | 4 | 689 | |||
BC111773.1 | - | 4 | 673 | 40034462 |