Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITFG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305002.1 | NP_001291931.1 | 3 | 247 | ||
NM_030790.4 | NP_110417.2 | 3 | 524 | |||
GenBank mRNA | AF212247.1 | - | 3 | 432 | ||
AF503339.1 | - | 3 | 314 | |||
AK027596.1 | - | 3 | 315 | |||
AK301908.1 | - | 3 | 199 | |||
AK309475.1 | - | 2 | 42 | |||
BC006321.2 | - | 3 | 297 | 4137934 | ||
BC024162.1 | - | 3 | 358 | 4046297 |