Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077484.1 | NP_001070952.1 | 3 | 827 | ||
NM_001278387.1 | NP_001265316.1 | 4 | 1289 | |||
NM_001278388.1 | NP_001265317.1 | 2 | 1051 | |||
NM_001278389.1 | NP_001265318.1 | 3 | 316 | |||
NM_001278390.1 | NP_001265319.1 | 3 | 1029 | |||
NM_030674.3 | NP_109599.3 | 3 | 1166 | |||
XM_011538784.2 | XP_011537086.1 | 3 | 551 | |||
XM_011538786.2 | XP_011537088.1 | 3 | 551 | |||
XM_011538787.2 | XP_011537089.1 | 3 | 551 | |||
XM_017019990.1 | XP_016875479.1 | 3 | 781 | |||
GenBank mRNA | AF247166.1 | - | 3 | 512 | ||
AF271070.1 | - | 3 | 410 | |||
AK027825.1 | - | 3 | 316 | |||
AK074851.1 | - | 1 | 27 | |||
AK074949.1 | - | 1 | 74 | |||
AK225639.1 | - | 3 | 404 | |||
AK225778.1 | - | 3 | 426 | |||
AK309410.1 | - | 3 | 743 | |||
AK313731.1 | - | 1 | 117 | |||
BC010620.1 | - | 3 | 373 | 3871101 | ||
BX647547.1 | - | 4 | 861 | |||
BX648495.1 | - | 3 | 754 | |||
BX648676.1 | - | 2 | 621 |