Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271723.1 | NP_001258652.1 | 13 | 1837 | ||
NM_030793.4 | NP_110420.3 | 13 | 1837 | |||
XM_005268513.1 | XP_005268570.1 | 13 | 1826 | |||
XM_006714797.1 | XP_006714860.1 | 13 | 1826 | |||
XM_011537684.2 | XP_011535986.1 | 13 | 1879 | |||
XM_017009899.1 | XP_016865388.1 | 14 | 1974 | |||
XM_017009900.1 | XP_016865389.1 | 14 | 2038 | |||
XM_017009901.1 | XP_016865390.1 | 13 | 1891 | |||
XM_017009902.1 | XP_016865391.1 | 13 | 1879 | |||
GenBank mRNA | AF251055.1 | - | 6 | 775 | ||
AK024024.1 | - | 13 | 1758 | |||
AK308414.1 | - | 13 | 1786 | |||
AL832841.1 | - | 3 | 304 | |||
BC005849.2 | - | 3 | 411 | |||
BC005873.2 | - | 3 | 411 | |||
BC033454.1 | - | 13 | 1770 | 4510274 | ||
BC050424.1 | - | 12 | 1742 | 6472693 |