Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STMN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283053.1 | NP_001269982.1 | 4 | 309 | ||
NM_030795.3 | NP_110422.2 | 4 | 309 | |||
XM_005273652.4 | XP_005273709.1 | 4 | 735 | |||
GenBank mRNA | AK294023.1 | - | 4 | 274 | ||
AK297415.1 | - | 4 | 290 | |||
BC011520.1 | - | 4 | 256 | 4157389 |