Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030918.5 | NP_112180.4 | 3 | 619 | ||
XM_005245509.2 | XP_005245566.1 | 3 | 741 | |||
XM_005245510.3 | XP_005245567.1 | 3 | 351 | |||
XM_005245511.4 | XP_005245568.1 | 3 | 647 | |||
XM_011510024.2 | XP_011508326.1 | 2 | 462 | |||
XM_011510025.2 | XP_011508327.1 | 2 | 320 | |||
XM_011510026.2 | XP_011508328.1 | 3 | 715 | |||
XM_017002417.1 | XP_016857906.1 | 2 | 320 | |||
GenBank mRNA | AB007957.2 | - | 3 | 588 | ||
AY044866.1 | - | 2 | 324 | |||
BC012184.1 | - | 3 | 609 | |||
BC071825.1 | - | 2 | 82 | |||
BC100998.1 | - | 3 | 619 | 40006277 | ||
BC100999.1 | - | 3 | 617 | 40006279 | ||
BC101000.1 | - | 3 | 619 | 40006281 | ||
BC101822.1 | - | 3 | 654 | 8069328 | ||
BC101824.1 | - | 3 | 654 | 8069330 | ||
BC107862.1 | - | 1 | 75 | |||
BM560557.1 | - | 620 |