Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITM2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012514.2 | NP_001012532.1 | 2 | 333 | ||
NM_001012516.2 | NP_001012534.1 | 3 | 419 | |||
NM_001287240.1 | NP_001274169.1 | 3 | 410 | |||
NM_001287241.1 | NP_001274170.1 | 4 | 606 | |||
NM_030926.5 | NP_112188.1 | 3 | 474 | |||
GenBank mRNA | AB003629.1 | - | 3 | 343 | ||
AF271781.1 | - | 3 | 459 | |||
AF272043.1 | - | 3 | 456 | |||
AK056321.1 | - | 2 | 280 | |||
AK075361.1 | - | 3 | 439 | |||
AK090975.1 | - | 3 | 385 | |||
AK222697.1 | - | 3 | 421 | |||
AK295503.1 | - | 3 | 387 | |||
AK303199.1 | - | 3 | 439 | |||
AK307702.1 | - | 2 | 242 | |||
AL136603.1 | - | 3 | 415 | |||
AL713651.1 | - | 3 | 398 | |||
AY049777.1 | - | 3 | 360 | |||
BC002424.1 | - | 3 | 401 | 3162430 | ||
BC025742.1 | - | 3 | 389 | 5224043 | ||
BC050668.1 | - | 2 | 268 | 5759920 | ||
BC098563.1 | - | 3 | 387 | 30558635 | ||
BM842614.1 | - | 3 | 463 | |||
BX444331.2 | - | 3 | 409 | |||
CR457208.1 | - | 3 | 343 | |||
DA138409.1 | - | 4 | 443 | |||
DA182471.1 | - | 3 | 476 |