Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC93B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030930.3 | NP_112192.2 | 6 | 832 | ||
XM_011545290.1 | XP_011543592.1 | 4 | 442 | |||
XM_011545291.2 | XP_011543593.1 | 5 | 724 | |||
GenBank mRNA | AJ271326.1 | - | 6 | 791 | ||
AJ422142.1 | - | 6 | 800 | |||
AY007125.1 | - | 5 | 619 | |||
BC025669.1 | - | 6 | 778 | |||
BC033623.1 | - | 6 | 830 | |||
BC092472.1 | - | 6 | 800 | 30348121 | ||
BC101568.1 | - | 6 | 776 | 8069074 | ||
BC105104.1 | - | 6 | 776 | 8144107 |