Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RNF146 SIRNA Assays ›
Gene Symbol:
RNF146
Gene Name:
ring finger protein 146
Gene Aliases:
-
Chromosome Location:
Chr.6: 127266340 - 127289893 on Build GRCh38
UniGene ID:
Hs.267120
Species:
Human
siRNA ID
HSS130076
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RNF146
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242844.1
NP_001229773.1
5
968
NM_001242845.1
NP_001229774.1
5
817
NM_001242846.1
NP_001229775.1
5
730
NM_001242847.1
NP_001229776.1
4
859
NM_001242848.1
NP_001229777.1
4
589
NM_001242849.1
NP_001229778.1
3
731
NM_001242850.1
NP_001229779.1
3
461
NM_001242851.1
NP_001229780.1
3
401
NM_001242852.1
NP_001229781.1
5
948
NM_030963.3
NP_112225.2
5
698
XM_006715571.3
XP_006715634.1
5
1398
XM_011536161.2
XP_011534463.1
6
1285
XM_011536162.2
XP_011534464.1
6
1337
XM_011536163.2
XP_011534465.1
6
1317
XM_011536164.2
XP_011534466.1
5
1176
XM_017011336.1
XP_016866825.1
3
1078
XM_017011337.1
XP_016866826.1
5
4319
XM_017011338.1
XP_016866827.1
7
1426
XM_017011339.1
XP_016866828.1
6
1455
XM_017011340.1
XP_016866829.1
6
1507
XM_017011341.1
XP_016866830.1
4
1206
XM_017011342.1
XP_016866831.1
5
1347
XM_017011343.1
XP_016866832.1
6
1456
GenBank mRNA
AJ315122.1
-
1
263
AJ420549.1
-
2
333
AK027436.1
-
2
402
AK027558.1
-
3
450
AK027776.1
-
1
273
AL136829.1
-
5
661
BC008235.1
-
2
350
3868592
BI771613.1
-
2
400
CN388414.1
-
3
541
CR533514.1
-
1
262
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
Wnt signaling pathway
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
protein autoubiquitination
protein K48-linked ubiquitination
positive regulation of canonical Wnt signaling pathway
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity
poly-ADP-D-ribose binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline