Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127496.1 | NP_001120968.1 | 2 | 226 | ||
NM_001293289.1 | NP_001280218.1 | 3 | 604 | |||
NM_001293290.1 | NP_001280219.1 | 2 | 542 | |||
NM_030964.3 | NP_112226.2 | 3 | 277 | |||
XM_011537685.2 | XP_011535987.1 | 3 | 1004 | |||
XM_017009910.1 | XP_016865399.1 | 2 | 1270 | |||
GenBank mRNA | AF227516.1 | - | 3 | 223 | ||
AK096464.1 | - | 2 | 211 | |||
AK226147.1 | - | 1 | 649 | |||
BC125095.1 | - | 2 | 75 | 40118267 | ||
BC125096.1 | - | 2 | 75 | 40118268 | ||
DA183391.1 | - | 2 | 226 |