Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFXN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030971.3 | NP_112233.2 | 3 | 561 | ||
GenBank mRNA | AK074707.1 | - | 2 | 174 | ||
AK299405.1 | - | 2 | 168 | |||
AL701236.1 | - | 3 | 567 | |||
BC000124.1 | - | 2 | 167 | 3352015 | ||
BG775086.1 | - | 1 | 37 | |||
BI819328.1 | - | 3 | 498 |