Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031212.3 | NP_112489.3 | 4 | 1005 | ||
XM_005270208.1 | XP_005270265.1 | 4 | 995 | |||
XM_005270211.4 | XP_005270268.1 | 4 | 609 | |||
XM_006718005.3 | XP_006718068.1 | 4 | 669 | |||
XM_011540239.2 | XP_011538541.1 | 4 | 891 | |||
XM_011540240.1 | XP_011538542.1 | 5 | 2141 | |||
XM_017016741.1 | XP_016872230.1 | 4 | 2077 | |||
XM_017016742.1 | XP_016872231.1 | 4 | 1898 | |||
GenBank mRNA | AF267854.1 | - | 1 | 692 | ||
AF327402.1 | - | 4 | 905 | |||
AF327403.1 | - | 4 | 1346 | |||
AJ303077.1 | - | 4 | 905 | |||
AJ303078.1 | - | 4 | 1346 | |||
AK056782.1 | - | 1 | 1045 | |||
AL831943.1 | - | 1 | 567 | |||
BC047312.1 | - | 4 | 952 | |||
BC058937.1 | - | 4 | 914 | 6139015 | ||
BC076399.1 | - | 4 | 945 | 5728224 | ||
BC094821.1 | - | 4 | 688 |