Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PITPNM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165966.1 | NP_001159438.1 | 14 | 1964 | ||
NM_031220.3 | NP_112497.2 | 15 | 2072 | |||
XM_011524015.2 | XP_011522317.1 | 15 | 2012 | |||
XM_011524016.2 | XP_011522318.1 | 15 | 2013 | |||
GenBank mRNA | AB209130.1 | - | 9 | 4435 | ||
AF334586.1 | - | 15 | 2069 | |||
AL389994.1 | - | 14 | 1795 | |||
BC020484.1 | - | 6 | 670 | |||
BC128583.1 | - | 15 | 2009 | 40128716 | ||
BC128584.1 | - | 15 | 2009 | 40128724 |