Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080554.2 | NP_001074023.1 | 6 | 1024 | ||
NM_001080555.2 | NP_001074024.1 | 6 | 900 | |||
NM_001206842.1 | NP_001193771.1 | 5 | 831 | |||
NM_001206843.1 | NP_001193772.1 | 5 | 723 | |||
NM_001206845.1 | NP_001193774.1 | 4 | 654 | |||
NM_031289.3 | NP_112579.2 | 5 | 931 | |||
NM_153823.3 | NP_722545.2 | 5 | 807 | |||
XM_005253493.2 | XP_005253550.1 | 6 | 954 | |||
XM_005253495.1 | XP_005253552.1 | 6 | 1078 | |||
XM_011520858.1 | XP_011519160.1 | 6 | 981 | |||
XM_011520859.1 | XP_011519161.1 | 6 | 898 | |||
XM_011520860.1 | XP_011519162.1 | 6 | 1105 | |||
XM_011520861.1 | XP_011519163.1 | 5 | 882 | |||
XM_011520862.1 | XP_011519164.1 | 6 | 1022 | |||
XM_011520863.1 | XP_011519165.1 | 5 | 1006 | |||
GenBank mRNA | AB593019.1 | - | 5 | 803 | ||
AB593020.1 | - | 5 | 679 | |||
AK027894.1 | - | 5 | 900 | |||
AK058045.1 | - | 6 | 973 | |||
AK075288.1 | - | 5 | 801 | |||
AK075311.1 | - | 4 | 496 | |||
AK075322.1 | - | 5 | 677 | |||
AY358513.1 | - | 5 | 785 | |||
BC001796.1 | - | 5 | 877 | 3353821 | ||
BC033854.1 | - | 5 | 774 | 5164943 | ||
BC112896.1 | - | 5 | 591 | 6145063 |