Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB33B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031296.2 | NP_112586.1 | 2 | 823 | ||
XM_011532299.1 | XP_011530601.1 | 3 | 919 | |||
GenBank mRNA | AF350420.1 | - | 2 | 545 | ||
AK313685.1 | - | 2 | 483 | |||
AL136904.1 | - | 2 | 823 | |||
BC036064.1 | - | 2 | 455 | 5288066 | ||
BC111977.1 | - | 2 | 475 | 8327445 |