Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031423.3 | NP_113611.2 | 14 | 1507 | ||
NM_145697.2 | NP_663735.2 | 14 | 1653 | |||
XM_011510036.1 | XP_011508338.1 | 14 | 1449 | |||
GenBank mRNA | AB050577.1 | - | 14 | 1568 | ||
AB050578.1 | - | 8 | 872 | |||
AF326731.1 | - | 14 | 1484 | |||
AK093348.1 | - | 14 | 1632 | |||
BC008489.1 | - | 14 | 1440 | 4134468 | ||
BC021171.2 | - | 8 | 922 |