Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300741.1 | NP_001287670.1 | 6 | 1226 | ||
NM_031438.3 | NP_113626.1 | 6 | 1280 | |||
XM_005272095.1 | XP_005272152.1 | 6 | 1265 | |||
XM_005272097.3 | XP_005272154.1 | 6 | 1226 | |||
GenBank mRNA | AK098066.1 | - | 6 | 1249 | ||
AK304010.1 | - | 6 | 1214 | |||
AL136592.1 | - | 6 | 1235 | |||
BC026748.1 | - | 3 | 302 | |||
BC041099.1 | - | 6 | 1264 | 5495144 |