Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031439.3 | NP_113627.1 | 2 | 1132 | ||
GenBank mRNA | AJ409320.1 | - | 2 | 991 | ||
AK055556.1 | - | 2 | 1070 | |||
AK223574.1 | - | 2 | 1070 | |||
BC004299.1 | - | 2 | 1059 | |||
BC071947.1 | - | 2 | 1058 | 6459570 | ||
BT006693.1 | - | 2 | 991 |