Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031457.1 | NP_113645.1 | 2 | 356 | ||
GenBank mRNA | AF237905.1 | - | 2 | 356 | ||
AF350504.1 | - | 2 | 198 | |||
AK310872.1 | - | 2 | 183 | |||
AK310884.1 | - | 2 | 348 | |||
AK310969.1 | - | 2 | 345 | |||
AK315901.1 | - | 2 | 318 | |||
BC022895.1 | - | 2 | 281 | 4691480 |