Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031457.1 | NP_113645.1 | 3 | 500 | ||
GenBank mRNA | AF237905.1 | - | 3 | 500 | ||
AF350504.1 | - | 3 | 342 | |||
AK308413.1 | - | 2 | 548 | |||
AK310872.1 | - | 4 | 517 | |||
AK310884.1 | - | 3 | 492 | |||
AK310969.1 | - | 3 | 589 | |||
AK315901.1 | - | 3 | 562 | |||
BC022895.1 | - | 3 | 425 | 4691480 |