Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS6KL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031464.4 | NP_113652.2 | 2 | 678 | ||
XM_011537206.2 | XP_011535508.1 | 3 | 906 | |||
XM_011537207.2 | XP_011535509.1 | 2 | 364 | |||
XM_011537211.2 | XP_011535513.1 | 3 | 938 | |||
XM_011537213.2 | XP_011535515.1 | 3 | 938 | |||
XM_017021680.1 | XP_016877169.1 | 2 | 326 | |||
XM_017021681.1 | XP_016877170.1 | 2 | 326 | |||
XM_017021682.1 | XP_016877171.1 | 2 | 316 | |||
XM_017021683.1 | XP_016877172.1 | 2 | 326 | |||
XM_017021684.1 | XP_016877173.1 | 2 | 326 | |||
XM_017021685.1 | XP_016877174.1 | 2 | 326 | |||
XM_017021686.1 | XP_016877175.1 | 2 | 326 | |||
XM_017021687.1 | XP_016877176.1 | 2 | 326 | |||
XM_017021688.1 | XP_016877177.1 | 2 | 326 | |||
XM_017021689.1 | XP_016877178.1 | 2 | 326 | |||
XM_017021690.1 | XP_016877179.1 | 2 | 326 | |||
XM_017021691.1 | XP_016877180.1 | 3 | 938 | |||
XM_017021692.1 | XP_016877181.1 | 2 | 326 | |||
GenBank mRNA | AK226137.1 | - | 2 | 277 | ||
AK299846.1 | - | 2 | 444 | |||
BC004540.2 | - | 2 | 410 | 3945902 |