Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017440.2 | NP_001017440.1 | 5 | 936 | ||
NM_031468.3 | NP_113656.2 | 6 | 926 | |||
XM_011516594.2 | XP_011514896.1 | 5 | 778 | |||
XM_011516595.1 | XP_011514897.1 | 5 | 526 | |||
XM_011516596.2 | XP_011514898.1 | 6 | 1421 | |||
XM_011516597.2 | XP_011514899.1 | 5 | 1354 | |||
XM_011516599.1 | XP_011514901.1 | 5 | 725 | |||
XM_017012675.1 | XP_016868164.1 | 7 | 1281 | |||
XM_017012676.1 | XP_016868165.1 | 6 | 1063 | |||
XM_017012677.1 | XP_016868166.1 | 5 | 576 | |||
XM_017012678.1 | XP_016868167.1 | 6 | 1038 | |||
XM_017012679.1 | XP_016868168.1 | 5 | 1391 | |||
XM_017012680.1 | XP_016868169.1 | 5 | 690 | |||
XM_017012681.1 | XP_016868170.1 | 5 | 553 | |||
XM_017012682.1 | XP_016868171.1 | 5 | 1103 | |||
XM_017012683.1 | XP_016868172.1 | 6 | 887 | |||
GenBank mRNA | AF282250.1 | - | 6 | 657 | ||
AK123862.1 | - | 5 | 471 | |||
AK313054.1 | - | 5 | 526 | |||
AY007302.1 | - | 5 | 584 | |||
AY726595.1 | - | 4 | 412 | |||
BC020200.1 | - | 5 | 708 | 3944302 |