Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIOK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031480.2 | NP_113668.2 | 9 | 971 | ||
NM_153005.1 | NP_694550.1 | 2 | 281 | |||
XM_005249446.3 | XP_005249503.1 | 9 | 1016 | |||
XM_011514933.2 | XP_011513235.2 | 9 | 933 | |||
GenBank mRNA | AF258661.1 | - | 2 | 281 | ||
AK054568.1 | - | 9 | 962 | |||
AK314467.1 | - | 9 | 1000 | |||
AL834277.1 | - | 8 | 670 | |||
BC006104.1 | - | 9 | 971 | 3537086 |