Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131028.1 | NP_001124500.1 | 6 | 798 | ||
NM_031482.4 | NP_113670.1 | 5 | 663 | |||
XM_005248610.4 | XP_005248667.1 | 5 | 1933 | |||
XM_005248611.4 | XP_005248668.1 | 5 | 507 | |||
XM_005248612.3 | XP_005248669.1 | 4 | 559 | |||
XM_011543660.2 | XP_011541962.1 | 4 | 363 | |||
XM_011543661.2 | XP_011541963.1 | 3 | 746 | |||
XM_017009944.1 | XP_016865433.1 | 5 | 694 | |||
GenBank mRNA | AK225969.1 | - | 4 | 555 | ||
AK315752.1 | - | 5 | 663 | |||
AL136912.1 | - | 5 | 531 | |||
BC029268.1 | - | 5 | 633 | 4521833 |