Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257137.2 | NP_001244066.1 | 15 | 1717 | ||
NM_001257138.2 | NP_001244067.1 | 13 | 1452 | |||
NM_001324197.1 | NP_001311126.1 | 15 | 1738 | |||
NM_001324198.1 | NP_001311127.1 | 14 | 1615 | |||
NM_031483.6 | NP_113671.3 | 14 | 1594 | |||
XM_005260570.1 | XP_005260627.1 | 13 | 1510 | |||
XM_011529079.1 | XP_011527381.1 | 13 | 1530 | |||
XM_017028089.1 | XP_016883578.1 | 14 | 1550 | |||
XM_017028090.1 | XP_016883579.1 | 15 | 1670 | |||
XM_017028091.1 | XP_016883580.1 | 13 | 1465 | |||
GenBank mRNA | AB056663.2 | - | 14 | 1504 | ||
AB209747.1 | - | 6 | 611 | |||
AF038564.1 | - | 9 | 1012 | |||
AF095745.1 | - | 12 | 1387 | |||
AK297809.1 | - | 14 | 1412 | |||
AK304090.1 | - | 13 | 1411 | |||
AK315212.1 | - | 14 | 1553 | |||
BC011571.1 | - | 15 | 1640 | 4560366 |