Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031485.3 | NP_113673.3 | 2 | 480 | ||
GenBank mRNA | AF337808.1 | - | 2 | 259 | ||
AK074676.1 | - | 2 | 448 | |||
AK131055.1 | - | 2 | 286 | |||
AK300225.1 | - | 2 | 403 | |||
AK315625.1 | - | 2 | 272 | |||
AL713729.1 | - | 2 | 255 | |||
BC002440.2 | - | 2 | 255 | 3347366 | ||
BC036281.1 | - | 2 | 231 | |||
BC047351.1 | - | 2 | 231 | |||
DA200417.1 | - | 2 | 481 | |||
DA714522.1 | - | 2 | 459 | |||
EF011614.1 | - | 2 | 253 |