Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031485.3 | NP_113673.3 | 3 | 684 | ||
GenBank mRNA | AF337808.1 | - | 3 | 463 | ||
AK074676.1 | - | 3 | 652 | |||
AK315625.1 | - | 3 | 476 | |||
AL713729.1 | - | 3 | 459 | |||
BC002440.2 | - | 3 | 459 | 3347366 | ||
BC036281.1 | - | 3 | 435 | |||
BC047351.1 | - | 3 | 435 | |||
EF011614.1 | - | 3 | 457 |