Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSD1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287191.1 | NP_001274120.1 | 1, 2 | 205 | ||
NM_001287192.1 | NP_001274121.1 | 1, 2 | 205 | |||
NM_031919.4 | NP_114125.1 | 1, 2 | 205 | |||
XM_005252257.3 | XP_005252314.1 | 1, 2 | 200 | |||
XM_005252260.3 | XP_005252317.1 | 1, 2 | 190 | |||
XM_017015183.1 | XP_016870672.1 | 1, 2 | 200 | |||
XM_017015187.1 | XP_016870676.1 | 1, 2 | 200 | |||
GenBank mRNA | AF088031.1 | - | 1, 2 | 194 | ||
AF316830.1 | - | 1, 2 | 68 | |||
AK296433.1 | - | 1, 2 | 116 | |||
AK299491.1 | - | 1, 2 | 116 | |||
BP229804.1 | - | 1, 2 | 205 | |||
BP365251.1 | - | 1, 2 | 195 |