Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMTC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193451.1 | NP_001180380.1 | 14 | 2126 | ||
NM_175861.3 | NP_787057.2 | 14 | 2217 | |||
XM_005253499.4 | XP_005253556.1 | 16 | 2378 | |||
XM_017020003.1 | XP_016875492.1 | 15 | 2211 | |||
XM_017020004.1 | XP_016875493.1 | 15 | 2171 | |||
XM_017020005.1 | XP_016875494.1 | 13 | 1984 | |||
XM_017020007.1 | XP_016875496.1 | 12 | 1404 | |||
GenBank mRNA | AK055962.1 | - | 12 | 1378 | ||
AK123619.1 | - | 10 | 1808 | |||
BC028716.2 | - | 3 | 407 | |||
BC039460.1 | - | 1 | 53 | |||
BC042083.1 | - | 14 | 2217 | 5749757 | ||
GQ865601.1 | - | 14 | 2126 |