Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142624.2 | NP_001136096.2 | 9, 10 | 757 | ||
NM_001144942.1 | NP_001138414.1 | 8, 9 | 1209 | |||
NM_001144943.1 | NP_001138415.1 | 9, 10 | 781 | |||
NM_001256276.1 | NP_001243205.1 | 8, 9 | 849 | |||
NM_001256277.1 | NP_001243206.1 | 9, 10 | 915 | |||
NM_001256278.1 | NP_001243207.1 | 7, 8 | 610 | |||
NM_031934.5 | NP_114140.4 | 8, 9 | 1233 | |||
GenBank mRNA | AF322067.1 | - | 8, 9 | 609 | ||
AJ277106.1 | - | 9, 10 | 820 | |||
AK027312.1 | - | 9, 10 | 814 | |||
AK074689.1 | - | 8, 9 | 1211 | |||
AK297850.1 | - | 9, 10 | 665 | |||
AK304633.1 | - | 8, 9 | 764 | |||
BC016841.1 | - | 8, 9 | 1067 | 3902651 | ||
BC066904.1 | - | 9, 10 | 830 | 5260472 | ||
BC091510.1 | - | 8, 9 | 775 | 30512024 | ||
BG749591.1 | - | 7, 8 | 536 | |||
BT006702.1 | - | 8, 9 | 585 | |||
CX788239.1 | - | 9, 10 | 674 |