Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031947.3 | NP_114153.1 | 1 | 669 | ||
GenBank mRNA | AF332005.1 | - | 1 | 471 | ||
AF378119.1 | - | 1 | 651 | |||
AY208943.1 | - | 1 | 579 | |||
BC069458.1 | - | 1 | 471 | 7262278 | ||
BC100935.2 | - | 1 | 471 | 40004045 | ||
BC100936.1 | - | 1 | 471 | 40004046 |