Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317781.1 | NP_001304710.1 | 14 | 2243 | ||
XM_005263267.4 | XP_005263324.1 | 14 | 2686 | |||
XM_011532302.2 | XP_011530604.1 | 16 | 2821 | |||
XM_011532303.2 | XP_011530605.1 | 16 | 2818 | |||
XM_011532304.2 | XP_011530606.1 | 15 | 2785 | |||
XM_011532305.2 | XP_011530607.1 | 15 | 2722 | |||
XM_011532306.2 | XP_011530608.1 | 16 | 2821 | |||
XM_017008680.1 | XP_016864169.1 | 15 | 2785 | |||
XM_017008681.1 | XP_016864170.1 | 15 | 2656 | |||
XM_017008682.1 | XP_016864171.1 | 13 | 2521 | |||
XM_017008683.1 | XP_016864172.1 | 13 | 1730 | |||
XM_017008684.1 | XP_016864173.1 | 15 | 2002 | |||
XM_017008685.1 | XP_016864174.1 | 9 | 1448 | |||
GenBank mRNA | AF113223.1 | - | 8 | 1128 | ||
AK026814.1 | - | 7 | 938 | |||
AK095661.1 | - | 11 | 1647 | |||
AY044656.1 | - | 7 | 1043 | |||
AY601647.1 | - | 14 | 2243 |