Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300761.2 | NP_001287690.1 | 12 | 1486 | ||
NM_001317806.1 | NP_001304735.1 | 11 | 1385 | |||
NM_031956.3 | NP_114162.2 | 11 | 1385 | |||
NR_133922.1 | 12 | 1539 | ||||
XM_005263270.1 | XP_005263327.1 | 12 | 1468 | |||
XM_006714335.1 | XP_006714398.1 | 12 | 1468 | |||
XM_006714336.1 | XP_006714399.1 | 12 | 1468 | |||
XM_006714339.2 | XP_006714402.1 | 11 | 1282 | |||
GenBank mRNA | AF345910.1 | - | 11 | 1336 | ||
AK093145.1 | - | 12 | 1432 | |||
AK225725.1 | - | 12 | 1486 | |||
AK301807.1 | - | 11 | 1311 | |||
AK314066.1 | - | 11 | 1381 | |||
BC024193.2 | - | 11 | 1326 | 4825866 | ||
EF432564.1 | - | 9 | 1154 |