Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM133 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032021.2 | NP_114410.1 | 1 | 581 | ||
GenBank mRNA | AF247167.1 | - | 1 | 581 | ||
BC093843.1 | - | 1 | 427 | 7939688 | ||
BC112050.1 | - | 1 | 427 | 8327518 |
Gene Symbol: ARHGAP42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011542615.2 | XP_011540917.1 | 24 | 8975 | ||
XM_011542616.2 | XP_011540918.1 | 24 | 6744 | |||
XM_011542617.2 | XP_011540919.1 | 25 | 6630 | |||
XM_017017237.1 | XP_016872726.1 | 24 | 6933 | |||
XM_017017238.1 | XP_016872727.1 | 25 | 9116 |