Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM2D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032027.2 | NP_114416.1 | 1, 2 | 456 | ||
NR_135160.1 | 1, 2 | 456 | ||||
NR_135161.1 | 1, 2 | 456 | ||||
NR_135162.1 | 1 | 456 | ||||
GenBank mRNA | AF353990.1 | - | 1, 2 | 456 | ||
AI168073.1 | - | 1, 2 | 150 | |||
AK311444.1 | - | 1, 2 | 102 | |||
AK311705.1 | - | 1, 2 | 103 | |||
BC029486.1 | - | 1, 2 | 174 | 5271098 | ||
BC048995.1 | - | 1, 2 | 172 | |||
BG700625.1 | - | 1, 2 | 171 | |||
BU664262.1 | - | 1 | 100 |