Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCALD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040624.1 | NP_001035714.1 | 6 | 525 | ||
NM_001040625.1 | NP_001035715.1 | 5 | 438 | |||
NM_001040626.1 | NP_001035716.1 | 5 | 438 | |||
NM_001040627.1 | NP_001035717.1 | 4 | 351 | |||
NM_001040628.1 | NP_001035718.1 | 6 | 494 | |||
NM_001040629.1 | NP_001035719.1 | 5 | 407 | |||
NM_001040630.1 | NP_001035720.1 | 4 | 591 | |||
NM_032041.2 | NP_114430.2 | 2 | 500 | |||
XM_011517332.2 | XP_011515634.1 | 2 | 289 | |||
XM_011517333.2 | XP_011515635.1 | 6 | 436 | |||
XM_011517334.2 | XP_011515636.1 | 3 | 320 | |||
XM_011517335.2 | XP_011515637.1 | 4 | 412 | |||
XM_017013901.1 | XP_016869390.1 | 5 | 628 | |||
XM_017013902.1 | XP_016869391.1 | 3 | 299 | |||
GenBank mRNA | AB209015.1 | - | 2 | 240 | ||
AF251061.1 | - | 4 | 279 | |||
AK055073.1 | - | 2 | 258 | |||
AK096515.1 | - | 6 | 450 | |||
AK314523.1 | - | 2 | 259 | |||
AL136612.1 | - | 2 | 251 | |||
BC036098.1 | - | 4 | 278 | 5274150 | ||
BC063428.1 | - | 5 | 424 | 5240958 | ||
DA185335.1 | - | 5 | 368 | |||
DA211806.1 | - | 2 | 500 | |||
DA215759.1 | - | 4 | 432 | |||
DA417037.1 | - | 4 | 351 | |||
DA776593.1 | - | 6 | 421 | |||
DA779293.1 | - | 5 | 334 |