Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM172A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032042.5 | NP_114431.2 | 2 | 251 | ||
NR_028080.1 | 2 | 251 | ||||
XM_005272103.3 | XP_005272160.1 | 2 | 217 | |||
XM_005272105.3 | XP_005272162.1 | 2 | 216 | |||
XM_005272106.3 | XP_005272163.1 | 2 | 217 | |||
XM_005272108.3 | XP_005272165.1 | 2 | 246 | |||
XM_005272109.4 | XP_005272166.1 | 2 | 217 | |||
XM_005272111.4 | XP_005272168.1 | 2 | 217 | |||
XM_006714717.3 | XP_006714780.1 | 2 | 243 | |||
XM_011543668.2 | XP_011541970.1 | 2 | 217 | |||
XM_011543670.2 | XP_011541972.1 | 2 | 217 | |||
XM_011543671.2 | XP_011541973.1 | 2 | 217 | |||
XM_011543672.2 | XP_011541974.1 | 1 | 22 | |||
XM_011543673.2 | XP_011541975.1 | 1 | 22 | |||
XM_011543674.2 | XP_011541976.1 | 2 | 217 | |||
XM_017009952.1 | XP_016865441.1 | 1 | 22 | |||
XM_017009954.1 | XP_016865443.1 | 2 | 217 | |||
XM_017009961.1 | XP_016865450.1 | 2 | 161 | |||
GenBank mRNA | AK295127.1 | - | 2 | 251 | ||
AK297471.1 | - | 1 | 13 | |||
AK312930.1 | - | 2 | 156 | |||
AL136630.1 | - | 2 | 90 | |||
BC020584.2 | - | 1 | 53 | 4398813 | ||
DC324758.1 | - | 2 | 251 | |||
DC374283.1 | - | 2 | 236 |