Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KREMEN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039570.2 | NP_001034659.2 | 6 | 987 | ||
NM_032045.4 | NP_114434.3 | 6 | 987 | |||
XM_011530429.2 | XP_011528731.1 | 6 | 947 | |||
XM_011530430.2 | XP_011528732.1 | 4 | 670 | |||
XM_011530431.2 | XP_011528733.1 | 4 | 2357 | |||
XM_011530432.2 | XP_011528734.1 | 5 | 936 | |||
XM_017028989.1 | XP_016884478.1 | 4 | 670 | |||
GenBank mRNA | AB059618.1 | - | 6 | 928 | ||
AK056425.1 | - | 6 | 947 | |||
BC063787.1 | - | 6 | 999 | 6315614 |