Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253861.1 | NP_001240790.1 | 4 | 280 | ||
NM_032117.3 | NP_115493.1 | 4 | 280 | |||
NR_045605.1 | 4 | 280 | ||||
XM_005263275.2 | XP_005263332.1 | 4 | 239 | |||
XM_017008688.1 | XP_016864177.1 | 4 | 637 | |||
GenBank mRNA | AK312172.1 | - | 4 | 239 | ||
AY028916.1 | - | 4 | 219 | |||
BC032142.2 | - | 4 | 234 | 5095005 | ||
BF975093.1 | - | 4 | 249 | |||
BG613292.1 | - | 4 | 242 | |||
BG758047.1 | - | 4 | 249 | |||
BM820228.1 | - | 4 | 280 | |||
BM833450.1 | - | 4 | 280 |