Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032117.3 | NP_115493.1 | 6 | 501 | ||
NR_045605.1 | 8 | 662 | ||||
XM_005263275.2 | XP_005263332.1 | 6 | 460 | |||
XM_017008688.1 | XP_016864177.1 | 6 | 858 | |||
GenBank mRNA | AK312172.1 | - | 6 | 460 | ||
AY028916.1 | - | 6 | 440 | |||
BC032142.2 | - | 6 | 455 | 5095005 | ||
BF975093.1 | - | 8 | 630 | |||
BG758047.1 | - | 8 | 631 | |||
BU533354.1 | - | 3 | 157 |