Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318765.1 | NP_001305694.1 | 5 | 782 | ||
NM_032124.4 | NP_115500.1 | 5 | 725 | |||
XM_005258371.4 | XP_005258428.1 | 5 | 745 | |||
XM_006722557.2 | XP_006722620.1 | 6 | 817 | |||
XM_011526227.2 | XP_011524529.1 | 6 | 801 | |||
XM_011526228.2 | XP_011524530.1 | 6 | 799 | |||
XM_017026039.1 | XP_016881528.1 | 6 | 838 | |||
XM_017026040.1 | XP_016881529.1 | 6 | 817 | |||
GenBank mRNA | AK054590.1 | - | 5 | 762 | ||
AK292098.1 | - | 5 | 725 | |||
AL136681.1 | - | 5 | 716 | |||
BC011894.2 | - | 5 | 746 | 3944181 | ||
BC033031.1 | - | 5 | 761 | 4793566 | ||
BC038100.2 | - | 5 | 725 | 5266439 | ||
CR533487.1 | - | 4 | 592 | |||
FJ224322.1 | - | 5 | 716 |