Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM160A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098794.1 | NP_001092264.1 | 3 | 746 | ||
NM_032127.3 | NP_115503.2 | 3 | 746 | |||
XM_006718343.2 | XP_006718406.1 | 3 | 668 | |||
XM_011520397.2 | XP_011518699.1 | 3 | 682 | |||
XM_011520398.2 | XP_011518700.1 | 3 | 727 | |||
XM_011520399.2 | XP_011518701.1 | 3 | 727 | |||
GenBank mRNA | AB051546.1 | - | 3 | 651 | ||
AK056468.1 | - | 3 | 698 | |||
AK291702.1 | - | 3 | 671 | |||
AL136723.1 | - | 3 | 680 | |||
BC030825.1 | - | 3 | 659 | 4811942 |