Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM186B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032130.2 | NP_115506.1 | 4 | 2143 | ||
NR_027450.1 | 4 | 2300 | ||||
XM_006719625.2 | XP_006719688.1 | 4 | 2132 | |||
XM_006719626.2 | XP_006719689.1 | 4 | 2132 | |||
XM_006719627.3 | XP_006719690.1 | 4 | 2132 | |||
XM_011538796.2 | XP_011537098.1 | 4 | 2132 | |||
XM_011538797.2 | XP_011537099.1 | 4 | 2132 | |||
XM_017020008.1 | XP_016875497.1 | 4 | 2383 | |||
GenBank mRNA | AK093242.1 | - | 4 | 2132 | ||
AK302702.1 | - | 4 | 2300 | |||
AL136748.1 | - | 4 | 2064 | |||
AL713673.1 | - | 1 | 809 | |||
BC035621.1 | - | 4 | 2101 | 5171752 |